Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

56790

Sigma-Aldrich

Sodium iminodiacetate dibasic monohydrate

≥95.0% (T)

Synonym(s):

Iminodiacetic acid disodium salt monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HN(CH2COONa)2 · H2O
CAS Number:
Molecular Weight:
195.08
Beilstein:
3790611
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥95.0% (T)

form

solid

SMILES string

[Na+].[Na+].[H]O[H].[O-]C(=O)CNCC([O-])=O

InChI

1S/C4H7NO4.2Na.H2O/c6-3(7)1-5-2-4(8)9;;;/h5H,1-2H2,(H,6,7)(H,8,9);;;1H2/q;2*+1;/p-2

InChI key

YSBGCHXLMBAKPV-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service