Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

557153

Sigma-Aldrich

4-(Bromomethyl)benzenesulfonyl chloride

95%

Synonym(s):

α-Bromo-p-toluenesulfonyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrCH2C6H4SO2Cl
CAS Number:
Molecular Weight:
269.54
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

solid

mp

71-75 °C (lit.)

SMILES string

ClS(=O)(=O)c1ccc(CBr)cc1

InChI

1S/C7H6BrClO2S/c8-5-6-1-3-7(4-2-6)12(9,10)11/h1-4H,5H2

InChI key

QXTQWYZHHMQSQH-UHFFFAOYSA-N

Application

4-(Bromomethyl)benzenesulfonyl chloride may be used to synthesize:
  • 4-bromomethyl-N-(6-methoxy-quinolin-8-yl)-benzenesulfonamide
  • benzene-sulfonamide derivatives, which are potent Chemokine Receptor Type 4 (CXCR4) inhibitors
  • imidazole derivatives having antifungal activity

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pierluigi Teolato et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 13(8), 2238-2245 (2006-12-13)
Silica nanoparticles (about 15 nm diameters), which contain a derivative of 6-methoxy-8-(p-toluensulfonamido)-quinoline (TSQ) as a Zn(II) fluorescent probe covalently linked to the silica network, were prepared and studied as Zn(II) fluorescent chemosensors. The systems selectively detect Zn(II) ions in water
Azizur Rehman et al.
Pakistan journal of pharmaceutical sciences, 32(3), 987-996 (2019-07-07)
Heterocyclic chemistry is an important field of organic chemistry due to therapeutic potential. The minor modification in the structure of poly-functional compounds has great effect on therapeutic ability. In the presented research work, substituted 1,3,4-oxadiazole derivatives, 8a-p, have been synthesized
Suazette Reid Mooring et al.
ChemMedChem, 8(4), 622-632 (2013-03-08)
The interaction of CXCR4 with CXCL12 (SDF-1) plays a critical role in cancer metastasis by facilitating the homing of tumor cells to metastatic sites. Based on our previously published work on CXCR4 antagonists, we have synthesized a series of aryl

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service