Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

55543

Sigma-Aldrich

4-Hydroxy-2-methoxybenzaldehyde

≥98.0% (HPLC)

Synonym(s):

4-Hydroxy-o-anisaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H8O3
CAS Number:
Molecular Weight:
152.15
Beilstein:
2045122
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0% (HPLC)

reaction suitability

reagent type: cross-linking reagent
reagent type: linker

mp

158-161 °C

functional group

carbonyl
hydroxyl

polymer architecture

functionality: heterobifunctional

SMILES string

OC1=CC(OC)=C(C([H])=O)C=C1

InChI

1S/C8H8O3/c1-11-8-4-7(10)3-2-6(8)5-9/h2-5,10H,1H3

InChI key

WBIZZNFQJPOKDK-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Mathias Dimde et al.
Biomaterials science, 5(11), 2328-2336 (2017-10-17)
In the present study, a pH sensitive nanogel platform for gene delivery was developed. The cationic nanogels based on dendritic polyglycerol (dPG) and low molecular weight polyethylenimine units were able to encapsulate siRNA during the manufacturing process. The thiol-Michael nanoprecipitation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service