Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

553352

Sigma-Aldrich

5-Bromo-2,3-dimethoxybenzaldehyde

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H2(OCH3)2CHO
CAS Number:
Molecular Weight:
245.07
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

81-84 °C (lit.)

SMILES string

COc1cc(Br)cc(C=O)c1OC

InChI

1S/C9H9BrO3/c1-12-8-4-7(10)3-6(5-11)9(8)13-2/h3-5H,1-2H3

InChI key

RVMWFOFQRYTRHZ-UHFFFAOYSA-N

General description

5-Bromo-2,3-dimethoxybenzaldehyde can be prepared by employing 5-bromo-2-hydroxy-3-methoxybenzaldehyde as a starting reagent.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Flavonoids syntheses. V. Synthesis of flavonoids with three hydroxy and four methoxy groups and their spectral properties.
Iinuma M, et al.
Chemical & Pharmaceutical Bulletin, 35(2), 660-667 (1987)
The cyclisation of benzylaminonitriles. Part 7. Regiospecific formation of methoxy-substituted isoquinolin-4-ones using methylthio activating groups.
Gavin JP and Waigh RD.
Journal of the Chemical Society. Perkin Transactions 1, 3, 503-508 (1990)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service