Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

538388

Sigma-Aldrich

3-Phenylbenzyl bromide

97%

Synonym(s):

3-Bromomethylbiphenyl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H11Br
CAS Number:
Molecular Weight:
247.13
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

57-61 °C

SMILES string

BrCc1cccc(c1)-c2ccccc2

InChI

1S/C13H11Br/c14-10-11-5-4-8-13(9-11)12-6-2-1-3-7-12/h1-9H,10H2

InChI key

RTFPTPXBTIUISM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3-Phenylbenzyl bromide may be used in the synthesis of the following amino alcohols:
  • NH-benzyl-(1R,2S)-norephedrine
  • NH-3-phenylbenzyl-(1R,2S)-norephedrine
  • (1R,2S)-2-(benzylamino)-1,2-diphenylethanol

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amino alcohol coordination in ruthenium (II)-catalysed asymmetric transfer hydrogenation of ketones
Petra DGI, et al.
European Journal of Inorganic Chemistry, 1999.12, 2335-2341 (1999)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service