Skip to Content
MilliporeSigma
All Photos(1)

Documents

536083

Sigma-Aldrich

3,5-Bis(trifluoromethyl)benzenethiol

97%

Synonym(s):

3,5-Bis(trifluoromethyl)thiophenol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CF3)2C6H3SH
CAS Number:
Molecular Weight:
246.17
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.442 (lit.)

bp

167 °C (lit.)

density

1.46 g/mL at 25 °C (lit.)

SMILES string

FC(F)(F)c1cc(S)cc(c1)C(F)(F)F

InChI

1S/C8H4F6S/c9-7(10,11)4-1-5(8(12,13)14)3-6(15)2-4/h1-3,15H

InChI key

KCAQWPZIMLLEAF-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

150.1 °F - closed cup

Flash Point(C)

65.6 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Suhailah S Al-Jameel et al.
World journal of microbiology & biotechnology, 34(4), 52-52 (2018-03-20)
Hay synthesis of a novel series of symmetrically tetra-substituted thiophenyl zinc(II)phthalocyanines (RS)4ZnPcs 4a-c was reported. Their novel 4-thiophenyl-phthalonitriles precursors 3(a-c) were synthesized from their substituted thiophenols 2(a-c). They were screened for their in-vitro antitumor activity on Human lung adenocarcinoma (A549)
Elzbieta Stepula et al.
ACS applied materials & interfaces, 12(29), 32321-32327 (2020-06-24)
There is an urgent clinical need for multicolor imaging of single cancer cells (no ensemble averaging) for identifying heterogenous expression of predictive biomarkers. Specifically, the comprehensive characterization of single disseminated tumor cells (sDTCs) responsible for metastatic relapse is the key

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service