Skip to Content
MilliporeSigma
All Photos(3)

Documents

526126

Sigma-Aldrich

4-(Chloromethyl)benzyl alcohol

99%

Synonym(s):

1-(Chloromethyl)-4-(hydroxymethyl)benzene, 4-(Chloromethyl)benzenemethanol, 4-Chloromethylbenzyl alcohol, 4-Hydroxymethylbenzyl chloride, [4-(Chloromethyl)phenyl]methanol, p-(Chloromethyl)benzyl alcohol, p-(Hydroxymethyl)benzyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH2C6H4CH2OH
CAS Number:
Molecular Weight:
156.61
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

mp

58-60 °C (lit.)

SMILES string

OCc1ccc(CCl)cc1

InChI

1S/C8H9ClO/c9-5-7-1-3-8(6-10)4-2-7/h1-4,10H,5-6H2

InChI key

OGALXJIOJZXBBP-UHFFFAOYSA-N

General description

4-(Chloromethyl)benzyl alcohol can be synthesized from 4-(chloromethyl)benzoyl chloride.

Application

4-(Chloromethyl)benzyl alcohol may be used in the synthesis of 4-(chloromethyl)benzyl acetate.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Functional RAFT agents for radical-controlled polymerization: Quantitative synthesis of trithiocarbonates containing functional groups as RAFT agents using equivalent amount of CS2.
Aoyagi N and Endo T.
Journal of Polymer Science Part A: Polymer Chemistry, 47(14), 3702-3709 (2009)
Fire properties of polystyrene-clay nanocomposites.
Zhu J, et al.
Chemistry of Materials, 13(10), 3774-3780 (2001)
Milada Šírová et al.
Journal of drug targeting, 25(9-10), 796-808 (2017-07-21)
Polymer carriers based on N-(2-hydroxypropyl)methacrylamide (HPMA) copolymers with incorporated organic nitrates as nitric oxide (NO) donors were designed with the aim to localise NO generation in solid tumours, thus highly increasing the enhanced permeability and retention (EPR) effect. The NO
Rodolfo M Moraes et al.
Polymers, 12(6) (2020-06-04)
Well-defined amphiphilic, biocompatible and partially biodegradable, thermo-responsive poly(N-vinylcaprolactam)-b-poly(ε-caprolactone) (PNVCL-b-PCL) block copolymers were synthesized by combining reversible addition-fragmentation chain transfer (RAFT) and ring-opening polymerizations (ROP). Poly(N-vinylcaprolactam) containing xanthate and hydroxyl end groups (X-PNVCL-OH) was first synthesized by RAFT/macromolecular design by the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service