Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

51835

Sigma-Aldrich

Sodium 1-heptanesulfonate monohydrate

≥96.0% (T)

Synonym(s):

1-Heptanesulfonic acid sodium salt monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)5CH2SO3Na · H2O
CAS Number:
Molecular Weight:
220.26
Beilstein:
3731762
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥96.0% (T)

form

solid

functional group

sulfonic acid

SMILES string

O.[Na+].CCCCCCCS([O-])(=O)=O

InChI

1S/C7H16O3S.Na.H2O/c1-2-3-4-5-6-7-11(8,9)10;;/h2-7H2,1H3,(H,8,9,10);;1H2/q;+1;/p-1

InChI key

XWZCREJRXRKIRQ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Sodium 1-heptanesulfonate monohydrate can be used as:
  • A catalyst to synthesize 2-aryl-1-arylmethyl-1H-benzo[d]imidazoles through the condensation reaction of o-phenylenediamine with various aromatic aldehydes.
  • An ion-pair reagent for HPLC-UV analysis.

Caution

may discolor on storage

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development and Validation of an HPLC Assay For Dual Detection of Gentamicin Sulfate and Leucine From a Novel Dry Powder For Inhalation.
Kuehl PJ, et al.
Journal of Analytical and Bioanalytical Techniques , 3(152), 2-2 (2012)
Visible-Light-Driven Photoreactions of [(bpy)2Ru (II) L]Cl2 in Aqueous Solutions (bpy= Bipyridine, L= 1,2-Bis (4-(4'-methyl)-2, 2'-bipyridyl) Ethene).
Zhang H, et al.
The Journal of Physical Chemistry A, 112(5), 808-817 (2008)
Martin Kögler et al.
Journal of medicinal chemistry, 54(13), 4847-4862 (2011-06-11)
A series of 5-substituted 2'-deoxyuridine monophosphate analogues has been synthesized and evaluated as potential inhibitors of mycobacterial ThyX, a novel flavin-dependent thymidylate synthase in Mycobacterium tuberculosis. A systematic SAR study led to the identification of compound 5a, displaying an IC(50)
Marisa Freitas et al.
Phytochemistry, 110, 91-103 (2015-01-03)
The intensification of agricultural productivity is an important challenge worldwide. However, environmental stressors can provide challenges to this intensification. The progressive occurrence of the cyanotoxins cylindrospermopsin (CYN) and microcystin-LR (MC-LR) as a potential consequence of eutrophication and climate change is
Md Iftekhar Ahmad et al.
Drug development and industrial pharmacy, 41(5), 791-800 (2014-04-04)
In this study, a dimple-shaped chitosan carrier was developed for delivering the antituberculosis drug ethambutol dihydrochloride (EDH) from a dry powder inhaler (DPI) to the lungs. Nanosized drug particles were prepared using nanospray drying. The microsized carrier was developed from

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service