Skip to Content
MilliporeSigma
All Photos(1)

Documents

516589

Sigma-Aldrich

Cyclopentyl isocyanate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C5H9NCO
CAS Number:
Molecular Weight:
111.14
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

2.83 psi ( 20 °C)

Assay

97%

refractive index

n20/D 1.447 (lit.)

bp

147 °C (lit.)

density

0.986 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

O=C=NC1CCCC1

InChI

1S/C6H9NO/c8-5-7-6-3-1-2-4-6/h6H,1-4H2

InChI key

CZALJDQHONFVFU-UHFFFAOYSA-N

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 3 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

97.0 °F - closed cup

Flash Point(C)

36.11 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Laura H Heitman et al.
Journal of medicinal chemistry, 52(7), 2036-2042 (2009-03-20)
The luteinizing hormone (LH) receptor plays an important role in fertility and certain cancers. The endogenous ligands human chorionic gonadotropin (hCG) and LH bind to the large N terminal domain of the receptor. We recently reported on the first radiolabeled
Synthesis and Antibacterial Activity of Urea and Thiourea Derivatives of Anacardic Acid Mixture Isolated from A Natural Product Cashew Nut Shell Liquid (CNSL).
N Subhakara R, et al.
International Journal of Organic Chemistry (2012)
Electron paramagnetic resonance of free radicals in an adamantane matrix. III. Isocyanatoalkyl and isothiocyanatoalkyl radicals.
Wood DE, et al.
Journal of the American Chemical Society, 93(17), 4145-4148 (1971)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service