Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

49733

Sigma-Aldrich

Glutaric acid monoethyl ester chloride

≥97.0% (NT)

Synonym(s):

Ethyl 4-(chloroformyl)butyrate, Ethyl 5-chloro-5-oxovalerate, Ethyl glutaryl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCO(CH2)3COOC2H5
CAS Number:
Molecular Weight:
178.61
Beilstein:
1099817
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97.0% (NT)

refractive index

n20/D 1.443

bp

68-70 °C/0.07 mmHg (lit.)

storage temp.

2-8°C

SMILES string

CCOC(=O)CCCC(Cl)=O

InChI

1S/C7H11ClO3/c1-2-11-7(10)5-3-4-6(8)9/h2-5H2,1H3

InChI key

KKJAQUGGQMCNJY-UHFFFAOYSA-N

General description

Glutaric acid monoethyl ester chloride (ethyl glutaryl chloride) is a higher acyl chloride homolog. Its reaction with calix[4]arene to form ester group-containing calix[4]arenes has been reported.

Application

Glutaric acid monoethyl ester chloride may be used as a reagent during the multi-step synthesis of lipidic aminoalcohol and diamine derivatives.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Esther del Olmo et al.
Bioorganic & medicinal chemistry letters, 16(23), 6091-6095 (2006-09-27)
Lymphoproliferation inhibition and cytotoxicity of a number of lipidic aminoacids, aminoalcohols and diamines were evaluated as a preliminary screening to select potential immunomodulators. The four most potent/less toxic compounds were submitted to delayed hypersensibility (DTH) assays to define the best
Synthesis of Several Diester Group-Containing Calix [4] arenes.
Nam KC, et al.
Bull. Korean Chem. Soc., 17(6), 502-506 (1996)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service