Skip to Content
MilliporeSigma
All Photos(2)

Documents

493651

Sigma-Aldrich

Bis(benzene)chromium(0)

97%

Synonym(s):

Dibenzenechromium

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H12Cr
CAS Number:
Molecular Weight:
208.22
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

reaction suitability

core: chromium
reagent type: catalyst

mp

260-265 °C (lit.)

SMILES string

[Cr].c1ccccc1.c2ccccc2

InChI

1S/2C6H6.Cr/c2*1-2-4-6-5-3-1;/h2*1-6H;

InChI key

HVURSIGIEONDKB-UHFFFAOYSA-N

Application

Catalyst for:
  • Olefin hydrogenation
  • Electron transfer chain catalysis mechanism

  • Pre-catalyst for the hydrosilation of ketones and aldehydes, for the dehydrocoupling of triphenylsilane with primary alcohols

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service