Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

491942

Sigma-Aldrich

Bromocyclopentane-d9

reagent grade, ≥98 atom % D, ≥99% (CP)

Synonym(s):

Cyclopentyl bromide-d9

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C5D9Br
CAS Number:
Molecular Weight:
158.08
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.12

grade

reagent grade

Quality Level

isotopic purity

≥98 atom % D

Assay

≥99% (CP)

refractive index

n20/D 1.488 (lit.)

bp

137-139 °C (lit.)

density

1.473 g/mL at 25 °C (lit.)

application(s)

pharmaceutical

mass shift

M+9

SMILES string

[2H]C1([2H])C([2H])([2H])C([2H])([2H])C([2H])(Br)C1([2H])[2H]

InChI

1S/C5H9Br/c6-5-3-1-2-4-5/h5H,1-4H2/i1D2,2D2,3D2,4D2,5D

InChI key

BRTFVKHPEHKBQF-UHUJFCCWSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

107.6 °F - closed cup

Flash Point(C)

42.00 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service