Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

491446

Sigma-Aldrich

2-Naphthalene-d7-sulfonic acid hydrate

98 atom % D, 95% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10D7SO3H · xH2O
Molecular Weight:
215.28 (anhydrous basis)
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

isotopic purity

98 atom % D

Assay

95% (CP)

form

solid

mp

117-121 °C (lit.)

mass shift

M+7

SMILES string

O.[2H]c1c([2H])c([2H])c2c([2H])c(c([2H])c([2H])c2c1[2H])S(O)(=O)=O

InChI

1S/C10H8O3S.H2O/c11-14(12,13)10-6-5-8-3-1-2-4-9(8)7-10;/h1-7H,(H,11,12,13);1H2/i1D,2D,3D,4D,5D,6D,7D;

InChI key

HKGOFWIZZIYCOS-VEFZIWIXSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Resp. Sens. 1 - Skin Corr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service