Skip to Content
MilliporeSigma
All Photos(1)

Documents

490032

Sigma-Aldrich

L-Glutamine-15N2

98 atom % 15N

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H215NCOCH2CH2CH(15NH2)CO2H
CAS Number:
Molecular Weight:
148.13
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

98 atom % 15N

form

solid

optical activity

[α]25/D +33.0°, c = 2 in 5 M HCl

technique(s)

bio NMR: suitable

mp

185 °C (dec.) (lit.)

mass shift

M+2

SMILES string

[15NH2][C@@H](CCC([15NH2])=O)C(O)=O

InChI

1S/C5H10N2O3/c6-3(5(9)10)1-2-4(7)8/h3H,1-2,6H2,(H2,7,8)(H,9,10)/t3-/m0/s1/i6+1,7+1

InChI key

ZDXPYRJPNDTMRX-QLUYOLLXSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Min Yuan et al.
Nature protocols, 14(2), 313-330 (2019-01-27)
Targeted tandem mass spectrometry (LC-MS/MS) has been extremely useful for profiling small molecules extracted from biological sources, such as cells, bodily fluids and tissues. Here, we present a protocol for analysing incorporation of the non-radioactive stable isotopes carbon-13 (13C) and
M Zaeem Cader et al.
Cell, 180(2), 278-295 (2020-01-25)
Mutations in FAMIN cause arthritis and inflammatory bowel disease in early childhood, and a common genetic variant increases the risk for Crohn's disease and leprosy. We developed an unbiased liquid chromatography-mass spectrometry screen for enzymatic activity of this orphan protein.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service