Skip to Content
MilliporeSigma
All Photos(1)

Documents

488364

Sigma-Aldrich

Succinic acid-2,3-13C2

99 atom % 13C

Synonym(s):

Butanedioic acid-2,3-13C2

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO2C13CH213CH2CO2H
CAS Number:
Molecular Weight:
120.07
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

99 atom % 13C

form

solid

bp

235 °C

mp

187-190 °C (lit.)

mass shift

M+2

SMILES string

OC(=O)[13CH2][13CH2]C(O)=O

InChI

1S/C4H6O4/c5-3(6)1-2-4(7)8/h1-2H2,(H,5,6)(H,7,8)/i1+1,2+1

InChI key

KDYFGRWQOYBRFD-ZDOIIHCHSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Articles

Isotopic Labeling for NMR Spectroscopy of Biological Solids;

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service