Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

47998

Sigma-Aldrich

Fmoc-8-Aoc-OH

≥98.0% (HPLC)

Synonym(s):

8-(Fmoc-amino)caprylic acid, 8-(Fmoc-amino)octanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H27NO4
CAS Number:
Molecular Weight:
381.46
Beilstein:
8286497
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

Assay

≥98.0% (HPLC)

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

2-8°C

SMILES string

OC(=O)CCCCCCCNC(=O)OCC1c2ccccc2-c3ccccc13

InChI

1S/C23H27NO4/c25-22(26)14-4-2-1-3-9-15-24-23(27)28-16-21-19-12-7-5-10-17(19)18-11-6-8-13-20(18)21/h5-8,10-13,21H,1-4,9,14-16H2,(H,24,27)(H,25,26)

InChI key

QZQXRZXYWVQWAY-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mathias Dyrberg Loft et al.
Amino acids, 49(6), 1089-1100 (2017-03-21)
The urokinase-type plasminogen activator receptor (uPAR) is overexpressed in several cancers including glioblastoma (GBM) and is an established biomarker for metastatic potential. The uPAR-targeting peptide AE105-NH

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service