Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

479136

Sigma-Aldrich

Poly(allylamine) solution

20 wt. % in H2O

Synonym(s):

PAH

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH2CH(CH2NH2)]n
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

concentration

20 wt. % in H2O

refractive index

n20/D 1.383

viscosity

~50 cP(25 °C)(lit.)

density

1.02 g/mL at 25 °C

SMILES string

NCC=C

InChI

1S/C3H7N/c1-2-3-4/h2H,1,3-4H2

InChI key

VVJKKWFAADXIJK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Soluble in water, lower alcohols, ethylene glycol and formamide.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xin Gao et al.
International journal of nanomedicine, 7, 4037-4051 (2012-08-14)
We designed dual-functional nanoparticles for in vivo application using a modified electrostatic and covalent layer-by-layer assembly strategy to address the challenge of assessment and treatment of hormone-refractory prostate cancer. Core-shell nanoparticles were formulated by integrating three distinct functional components, ie
Ilaria E Palamà et al.
Biotechnology and bioengineering, 110(2), 586-596 (2012-08-14)
Alignment of skeletal myoblasts is considered a critical step during myotube formation. The C2C12 cell line is frequently used as a model of skeletal muscle differentiation that can be induced by lowering the serum concentration in standard culture flasks. In
Magdalena Oćwieja et al.
Langmuir : the ACS journal of surfaces and colloids, 29(11), 3546-3555 (2013-01-17)
Systematic studies of silver particle deposition kinetics under diffusion transport on poly(allylamine hydrochloride) (PAH) modified mica were carried out. Monolayer coverage, quantitatively determined by AFM and SEM, was regulated within broad limits by adjusting the deposition time and the ionic
Gengtao Fu et al.
Nanoscale, 5(17), 8007-8014 (2013-07-19)
This paper reports a facile, water-based and one-pot synthesis of tetrahedral Pd nanocrystals (Pd-TNPs) with high yield and good size monodispersity supported on reduced graphene oxide (RGO) nanosheets via a co-chemical reduction method. The key synthetic strategy employed a positively
L Minati et al.
International journal of pharmaceutics, 438(1-2), 45-52 (2012-09-11)
Gold nanoparticles functionalized with doxorubicin and stabilized with multilayers of degradable polyelectrolyte were allowed to age in aqueous medium in vitro in order to show the possibility of drug release in cellular environment. The chemico-physical characteristics of the nanoparticles are

Articles

We present an article that discusses two applications in particular; first, using these layers as polyelectrolyte membranes to control permeability.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service