Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

474193

Sigma-Aldrich

2,2,3,3,3-Pentafluoropropyl methacrylate

97%, contains 100 ppm 4-tert-butylcatechol as inhibitor

Synonym(s):

1,1-Dihydroperfluoropropyl methacrylate, 1H ,1H -Pentafluoropropyl methacrylate, 2,2,3,3,3-Pentafluoropropyl methacrylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=C(CH3)CO2CH2CF2CF3
CAS Number:
Molecular Weight:
218.12
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

contains

100 ppm 4-tert-butylcatechol as inhibitor

refractive index

n20/D 1.347 (lit.)

bp

55 °C/100 mmHg (lit.)

density

1.277 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CC(=C)C(=O)OCC(F)(F)C(F)(F)F

InChI

1S/C7H7F5O2/c1-4(2)5(13)14-3-6(8,9)7(10,11)12/h1,3H2,2H3

InChI key

CLISWDZSTWQFNX-UHFFFAOYSA-N

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

84.2 °F - closed cup

Flash Point(C)

29 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lucas Narciso Meirelles et al.
Journal of separation science, 41(19), 3669-3676 (2018-08-01)
Organo-silica monolithic capillary columns were prepared using thermally initiated polymerization of mixtures containing 3-(methacryloyloxy)propyltrimethoxysilane as the silicon-containing monomer, an aqueous acid catalyst, ethylene dimethacrylate as the cross-linker, a functional monomer selected from the group comprising octadecyl methacrylate, trifluoroethyl methacrylate, pentafluoropropyl

Articles

Polymeric materials, with unique refractive index properties, find applications in photonics and optical coatings for various devices.

Polymeric materials, with unique refractive index properties, find applications in photonics and optical coatings for various devices.

Polymeric materials, with unique refractive index properties, find applications in photonics and optical coatings for various devices.

Polymeric materials, with unique refractive index properties, find applications in photonics and optical coatings for various devices.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service