Skip to Content
MilliporeSigma
All Photos(1)

Documents

474169

Sigma-Aldrich

Glycidyl 2,2,3,3,4,4,5,5-octafluoropentyl ether

96%

Synonym(s):

Glycidyl 2,2,3,3,4,4,5,5-octafluoropentyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H8F8O2
CAS Number:
Molecular Weight:
288.14
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

96%

refractive index

n20/D 1.353 (lit.)

bp

75-79 °C/4 mmHg (lit.)

density

1.509 g/mL at 25 °C (lit.)

SMILES string

FC(F)C(F)(F)C(F)(F)C(F)(F)COCC1CO1

InChI

1S/C8H8F8O2/c9-5(10)7(13,14)8(15,16)6(11,12)3-17-1-4-2-18-4/h4-5H,1-3H2

InChI key

NABHRPSATHTFNS-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

224.6 °F - closed cup

Flash Point(C)

107.00 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service