Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

472670

Sigma-Aldrich

9,10-Difluoro-2,3-dihydro-3-methyl-7-oxo-7H-pyrido[1,2,3-de]-1,4-benzoxazine-6-carboxylic acid

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H9F2NO4
CAS Number:
Molecular Weight:
281.21
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

320-322 °C (lit.)

SMILES string

CC1COc2c(F)c(F)cc3C(=O)C(=CN1c23)C(O)=O

InChI

1S/C13H9F2NO4/c1-5-4-20-12-9(15)8(14)2-6-10(12)16(5)3-7(11(6)17)13(18)19/h2-3,5H,4H2,1H3,(H,18,19)

InChI key

NVKWWNNJFKZNJO-UHFFFAOYSA-N

Gene Information

human ... CSNK2A1(1457)

Looking for similar products? Visit Product Comparison Guide

General description

(S)-(-)-9,10-Difluoro-2,3-dihydro-3-methyl-7-oxo-7H-pyrido[1,2,3-de]-1,4-benzoxazine-6-carboxylic acid may be used in the preparation of the following:
  • kynurenine aminotransferase II (KAT II) inhibitor, [(S)-(-)-9-(4-aminopiperazine-1-yl)-8-fluoro-3-methyl-6-oxo-2,3,5,6-tetrahydro-4H-1-oxa-3a-aza-phenalene-5-carboxylic acid]
  • fused-ring derivatives
  • N-desmethyllevofloxacin

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

7-Piperazinylquinolones with methylene-bridged nitrofuran scaffold as new antibacterial agents.
Emami S, et al.
Medicinal Chemistry Research, 22(12), 5940-5947 (2013)
On the relationship between the two branches of the kynurenine pathway in the rat brain in vivo.
Amori L, et al.
Journal of Neurochemistry, 109(2), 316-325 (2009)
Muhammad Malik et al.
Antimicrobial agents and chemotherapy, 54(12), 5214-5221 (2010-09-22)
Quinolones rapidly kill bacteria by two mechanisms, one that requires protein synthesis and one that does not. The latter, which is measured as lethal action in the presence of the protein synthesis inhibitor chloramphenicol, is enhanced by N-1 cyclopropyl and
Andriy G Golub et al.
Journal of medicinal chemistry, 49(22), 6443-6450 (2006-10-27)
Due to the emerging role of protein kinase CK2 as a molecule that participates not only in the development of some cancers but also in viral infections and inflammatory failures, small organic inhibitors of CK2, besides application in scientific research

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service