Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

471585

Sigma-Aldrich

tert-Dodecylmercaptan

mixture of isomers, 98.5%

Synonym(s):

tert-Dodecanethiol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C12H25SH
CAS Number:
Molecular Weight:
202.40
Beilstein:
1738382
EC Number:
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.23

Quality Level

Assay

98.5%

bp

227-248 °C (lit.)

density

0.86 g/mL at 20 °C (lit.)

SMILES string

SC(C(C(C(C)C)(C)C)(C)C)(C)C

InChI

1S/C12H26S/c1-9(2)10(3,4)11(5,6)12(7,8)13/h9,13H,1-8H3

InChI key

YAJYJWXEWKRTPO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Data characterizing the biophysical and nitric oxide release properties of the tDodSNO-Styrene maleic anhydride nanoparticle SMA-tDodSNO: This study discusses the synthesis and characterization of SMA-tDodSNO nanoparticles using tert-dodecylmercaptan (H Alimoradi et al., 2018).

  • Cu2ZnSnS4-Ag2S Nanoscale p-n Heterostructures as Sensitizers for Photoelectrochemical Water Splitting: The research investigates the use of tert-dodecylmercaptan in the synthesis of CZTS-Ag2S heterostructures for photoelectrochemical water splitting (X Yu et al., 2015).

  • Enhanced performance of triboelectric nanogenerator based on polyamide-silver antimony sulfide nanofibers for energy harvesting: This study discusses the synthesis of nanofibers using tert-dodecylmercaptan for triboelectric nanogenerators (A Yar et al., 2021).

Legal Information

Product of Arkema Inc.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 1 - Skin Irrit. 2 - Skin Sens. 1B

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

203.0 °F - closed cup

Flash Point(C)

95 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service