Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

468258

Sigma-Aldrich

Poly(ethylene glycol) behenyl ether methacrylate solution

average Mn ~1,500, 50 wt. % in methacrylic acid/water, contains 1000 ppm MEHQ as stabilizer, 25% water

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=C(CH3)CO2(CH2CH2O)n(CH2)21CH3
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

mol wt

average Mn ~1,500

contains

1000 ppm MEHQ as stabilizer
25% water

concentration

50 wt. % in methacrylic acid/water

refractive index

n20/D 1.431

pH

3-4

viscosity

300 cP(25 °C)(lit.)

bp

95 °C

density

1.06 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

OCCO.CC(=C)C(O)=O.CCCCCCCCCCCCCCCCCCCCCCO

Application

Copolymerizable surfactant and associative thickener in acrylic latexes.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Resp. Sens. 1 - Skin Corr. 1A - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

235.4 °F

Flash Point(C)

113 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service