Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

463299

Sigma-Aldrich

Glycolic acid ethoxylate 4-tert-butylphenyl ether

average Mn ~380

Synonym(s):

Butoxynol-5 carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CC6H4(OCH2CH2)nOCH2CO2H, n~4
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

mol wt

average Mn ~380

refractive index

n20/D 1.49 (lit.)

density

1.12 g/mL at 25 °C (lit.)

SMILES string

OCCO.OCC(O)=O.CC(C)(C)c1ccc(O)cc1

InChI

1S/C10H14O.C2H4O3.C2H6O2/c1-10(2,3)8-4-6-9(11)7-5-8;3-1-2(4)5;3-1-2-4/h4-7,11H,1-3H3;3H,1H2,(H,4,5);3-4H,1-2H2

InChI key

OAGSUPHYBVNDLY-UHFFFAOYSA-N

Application

Anionic surfactant.
Emulsifier for metal-working fluids.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service