Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

463280

Sigma-Aldrich

Glycolic acid ethoxylate oleyl ether

average Mn ~700

Synonym(s):

Oleth-10 carboxylic acid, Oleth-3 carboxylic acid, Oleth-6 carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)xCH=CH(CH2)8(OCH2CH2)yOCH2CO2H, x = 5-7, y~2
CAS Number:
Molecular Weight:
901.47
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

mol wt

average Mn ~700

impurities

≤1.5% NaCl
6-10% water

refractive index

n20/D 1.461

density

0.99 g/mL at 25 °C

SMILES string

OCCO.OCC(O)=O.[H]\C(CCCCCC)=C(/[H])CCCCCCCCO.[H]\C(CCCCCCC)=C(/[H])CCCCCCCCO.[H]\C(CCCCCCCC)=C(/[H])CCCCCCCCO

InChI

1S/C18H36O.C17H34O.C16H32O.C2H4O3.C2H6O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19;1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18;1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17;3-1-2(4)5;3-1-2-4/h9-10,19H,2-8,11-18H2,1H3;8-9,18H,2-7,10-17H2,1H3;7-8,17H,2-6,9-16H2,1H3;3H,1H2,(H,4,5);3-4H,1-2H2/b10-9-;9-8-;8-7-;;

InChI key

UWYGJWXPMKLMHK-CDQAOSTISA-N

Looking for similar products? Visit Product Comparison Guide

Application

Anionic surfactant.
Emulsifier for metal-working fluids.

Features and Benefits

Insensitive to hard water.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service