Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

461059

Sigma-Aldrich

Benzyl-α,α-d2 alcohol

98 atom % D, 99% (CP)

Synonym(s):

a,a-Dideuteriobenzyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CD2OH
CAS Number:
Molecular Weight:
110.15
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

98 atom % D

Quality Level

Assay

99% (CP)

refractive index

n20/D 1.5388 (lit.)

bp

203-205 °C (lit.)

mp

-16--13 °C (lit.)

density

1.065 g/mL at 25 °C

mass shift

M+2

SMILES string

[2H]C([2H])(O)c1ccccc1

InChI

1S/C7H8O/c8-6-7-4-2-1-3-5-7/h1-5,8H,6H2/i6D2

InChI key

WVDDGKGOMKODPV-NCYHJHSESA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

213.8 °F - closed cup

Flash Point(C)

101 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service