Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

458449

Sigma-Aldrich

Titanium(IV) iodide

anhydrous, powder, 99.99% trace metals basis

Synonym(s):

Titanium tetraiodide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
TiI4
CAS Number:
Molecular Weight:
555.49
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
99.99% trace metals basis
grade:
anhydrous
form:
powder

grade

anhydrous

Quality Level

Assay

99.99% trace metals basis

form

powder

impurities

≤150.0 ppm Trace Metal Analysis

mp

150 °C (lit.)

density

4.3 g/mL at 25 °C (lit.)

SMILES string

I[Ti](I)(I)I

InChI

1S/4HI.Ti/h4*1H;/q;;;;+4/p-4

InChI key

NLLZTRMHNHVXJJ-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

Related Categories

Features and Benefits

Used in the CVD of titanium nitride films with optimized characteristics.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

K Sellschopp et al.
The Journal of chemical physics, 152(6), 064702-064702 (2020-02-18)
The ability to synthesize nanoparticles (NPs), here TiO2, of different shapes in a controlled and reproducible way is of high significance for a wide range of fields including catalysis and materials design. Different NP shapes exhibit variations of emerging facets

Articles

Titanium dioxide applications: Semiconducting material characteristics and diverse functionalities.

Titanium dioxide applications: Semiconducting material characteristics and diverse functionalities.

Titanium dioxide applications: Semiconducting material characteristics and diverse functionalities.

Titanium dioxide applications: Semiconducting material characteristics and diverse functionalities.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service