Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

455687

Sigma-Aldrich

Propene-d6

99 atom % D

Synonym(s):

Propylene-d6

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CD3CD=CD2
CAS Number:
Molecular Weight:
48.12
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

99 atom % D

Assay

99% (CP)

bp

−47.7 °C (lit.)

mp

−185 °C (lit.)

mass shift

M+6

SMILES string

[2H]\C([2H])=C(/[2H])C([2H])([2H])[2H]

InChI

1S/C3H6/c1-3-2/h3H,1H2,2H3/i1D2,2D3,3D

InChI key

QQONPFPTGQHPMA-QNYGJALNSA-N

Packaging

1 L quantity is packaged in a 450 mL carbon steel lecture bottle with brass CGA 110/180 valve. Nominal gas pressure at 21ºC is 20 psig for 1 L. This pressure is slightly above atmospheric pressure. Care must be taken when extracting this product from the cylinder.
All gas volumes are quoted at Standard Temperature and Pressure (STP) and not at Normal Temperature and Pressure (NTP). One STP Liter is 7.7% more gas than one NTP Liter.
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Recommended products

Control valve Z146943 (brass, 1/4" hose outlet) or Z187224 (SS, 1/4" compression fitting outlet)

Pictograms

FlameGas cylinder

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Gas 1 - Press. Gas Liquefied gas

Storage Class Code

2A - Gases

WGK

nwg

Flash Point(F)

-162.4 °F - closed cup

Flash Point(C)

-108 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service