Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

441864

Sigma-Aldrich

Glycerol ethoxylate

average Mn 1,000

Synonym(s):

Polyethylene glycol, Polyoxyethylenetriol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO(CH2CH2O)nCH[CH2(OCH2CH2)nOH]2
CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

product name

Glycerol ethoxylate, average Mn ~1,000

form

viscous liquid

mol wt

average Mn ~1,000
degree of polymerization 20.4

refractive index

n20/D 1.473 (lit.)

pH

5.5-7.5

viscosity

320 cSt(21 °C)(lit.)

bp

>200 °C (lit.)

hydroxyl value

160‑177

transition temp

softening point −6-(−4) °C

solubility

H2O: soluble

density

1.138 g/mL at 25 °C (lit.)

InChI

1S/C9H20O6/c10-1-4-13-7-9(15-6-3-12)8-14-5-2-11/h9-12H,1-8H2

InChI key

UCYLROVJSUACAD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Cross-linking agent for poly -esters, -urethanes, and -ethers.

Features and Benefits

Contributes flexibility in polymers.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service