Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

424684

Sigma-Aldrich

Phenethylmagnesium chloride solution

1.0 M in THF

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2CH2MgCl
CAS Number:
Molecular Weight:
164.92
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

reaction suitability

reaction type: Grignard Reaction

concentration

1.0 M in THF

bp

65 °C

density

0.954 g/mL at 25 °C

SMILES string

Cl[Mg]CCc1ccccc1

InChI

1S/C8H9.ClH.Mg/c1-2-8-6-4-3-5-7-8;;/h3-7H,1-2H2;1H;/q;;+1/p-1

InChI key

AZORNQXQALOFHH-UHFFFAOYSA-M

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

1.4 °F - closed cup

Flash Point(C)

-17 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oleg V Larionov et al.
Organic letters, 16(3), 864-867 (2014-01-15)
A one-step transformation of heterocyclic N-oxides to 2-alkyl-, aryl-, and alkenyl-substituted N-heterocycles is described. The success of this broad-scope methodology hinges on the combination of copper catalysis and activation by lithium fluoride or magnesium chloride. The utility of this method

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service