Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

418749

Sigma-Aldrich

Pentaerythritol propoxylate (5/4 PO/OH)

average Mn ~426

Synonym(s):

Pentaerythritol propoxylate (17/8 PO/OH)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C[CH2[OCH2CH(CH3)]nOH]4
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

form

viscous liquid

mol wt

average Mn ~426

refractive index

n20/D 1.464

bp

>300 °C (lit.)

density

1.05 g/mL at 25 °C (lit.)

SMILES string

CC(O)CO.OCC(CO)(CO)CO

InChI

1S/C17H36O8.C5H12O4/c18-5-1-9-22-13-17(14-23-10-2-6-19,15-24-11-3-7-20)16-25-12-4-8-21;6-1-5(2-7,3-8)4-9/h18-21H,1-16H2;6-9H,1-4H2

InChI key

PWIIWDSISSNGLT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pentaerythritol propoxylate (5/4 PO/OH) is a pentaerythriol based branched polymer that can be used as a crystallization agent to generate ordered, high resolution diffraction quality crystals.

Application

Pentaerythritol propoxylate may be used as a precipitant in reservoir solution for the crystallization of Phosphate Transporter(PiPT) and Aggregation Suppressing Protein A(AgsA). It may also acts as a reservoir solution for Phytosulfokine (PSK)- receptor PSKR- and somatic embryogenesis receptor-like kinases (SERKs) complex.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

377.6 °F - closed cup

Flash Point(C)

192 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Crystal structure of a eukaryotic phosphate transporter.
Pedersen BP, et al.
Nature, 496(7446), 533-533 (2013)
Multiple oligomeric structures of a bacterial small heat shock protein.
Mani N, et al.
Scientific Reports, 6(7446), 24019-24019 (2016)
Allosteric receptor activation by the plant peptide hormone phytosulfokine.
Wang J, et al.
Nature, 525(7568), 265-265 (2015)
Pentaerythritol propoxylate: a new crystallization agent and cryoprotectant induces crystal growth of 2-methylcitrate dehydratase.
Gulick A, et al.
Acta Crystallographica Section D, Biological Crystallography, 58(2), 306-309 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service