Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

413607

Sigma-Aldrich

Barium bromide

AnhydroBeads, −10 mesh, 99.999% trace metals basis

Synonym(s):

Barium dibromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BaBr2
CAS Number:
Molecular Weight:
297.14
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
99.999% trace metals basis
form:
beads

product line

AnhydroBeads

Quality Level

Assay

99.999% trace metals basis

form

beads

impurities

≤15.0 ppm Trace Metal Analysis

particle size

−10 mesh

SMILES string

Br[Ba]Br

InChI

1S/Ba.2BrH/h;2*1H/q+2;;/p-2

InChI key

NKQIMNKPSDEDMO-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Legal Information

AnhydroBeads is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service