Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

408069

Sigma-Aldrich

3-Methoxy-3-methyl-1-butanol

greener alternative

≥98%

Synonym(s):

3-Methoxy-3-methyl-1-butyl alcohol, 3-Methoxy-3-methylbutan-1-ol, 3-Methyl-3-(methyloxy)butan-1-ol, 3-Methyl-3-methoxy-1-hydroxybutane, 3-Methyl-3-methoxybutanol, 3-Methyl-3-methoxybutyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C(OCH3)CH2CH2OH
CAS Number:
Molecular Weight:
118.17
Beilstein:
1732882
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98%

form

liquid

greener alternative product characteristics

Safer Solvents and Auxiliaries
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

refractive index

n20/D 1.428 (lit.)

bp

173-175 °C (lit.)

density

0.926 g/mL at 20 °C (lit.)

greener alternative category

SMILES string

COC(C)(C)CCO

InChI

1S/C6H14O2/c1-6(2,8-3)4-5-7/h7H,4-5H2,1-3H3

InChI key

MFKRHJVUCZRDTF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3-Methoxy-3-methyl-1-butanol is widely used as solvent for paints, inks, and fragrances. Kinetics of the reaction of 3-methoxy-3-methyl-1-butanol with OH radicals using a relative rate method has been reported. It has been identified as oxidation products of dyes such as methyl yellow, methyl red and methyl orange.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product is considered as a multifunctional Chemical for a cleaner and Greener Environment and thus aligns with "Safer Solvents and Auxiliaries". Click here for more information.

Application

3-Methoxy-3-methyl-1-butanol may be employed as raw material for the production of industrial detergents.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

159.8 °F - closed cup

Flash Point(C)

71 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Degradation of organic dyes in water by electrical discharges.
Magureanu M, et al.
Plasma Chemistry and Plasma Processing, 27(5), 589-598 (2007)
Sara M Aschmann et al.
Environmental science & technology, 45(16), 6896-6901 (2011-07-22)
3-Methoxy-3-methyl-1-butanol [CH(3)OC(CH(3))(2)CH(2)CH(2)OH] is used as a solvent for paints, inks, and fragrances and as a raw material for the production of industrial detergents. A rate constant of (1.64 ± 0.18) × 10(-11) cm(3) molecule(-1) s(-1) for the reaction of 3-methoxy-3-methyl-1-butanol

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service