Skip to Content
MilliporeSigma
All Photos(3)

Documents

403237

Sigma-Aldrich

Dichlorobis(tricyclohexylphosphine)palladium(II)

95%

Synonym(s):

PdCl2[P(cy)3]2

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(C6H11)3P]2PdCl2
CAS Number:
Molecular Weight:
738.18
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

solid

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Cross Couplings
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst

mp

270 °C (dec.) (lit.)

SMILES string

Cl[Pd]Cl.C1CCC(CC1)P(C2CCCCC2)C3CCCCC3.C4CCC(CC4)P(C5CCCCC5)C6CCCCC6

InChI

1S/2C18H33P.2ClH.Pd/c2*1-4-10-16(11-5-1)19(17-12-6-2-7-13-17)18-14-8-3-9-15-18;;;/h2*16-18H,1-15H2;2*1H;/q;;;;+2/p-2

InChI key

VUYVXCJTTQJVKJ-UHFFFAOYSA-L

General description

Dichlorobis(tricyclohexylphosphine)palladium(II) is widely employed as catalyst precursor in various carbon-carbon and carbon-heteroatom bond forming reactions.

Application

Catalyst for C-C and C-N coupling reaction.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dichlorobis (tricyclohexylphosphine) palladium.
Carpentier JF, et al.
e-EROS Encyclopedia of Reagents for Organic Synthesis. (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service