Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

386391

Sigma-Aldrich

N-Ethylisopropylamine

98%

Synonym(s):

1-Methyldiethylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHNHC2H5
CAS Number:
Molecular Weight:
87.16
Beilstein:
505952
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.392 (lit.)

bp

71-73 °C (lit.)

density

0.72 g/mL at 25 °C (lit.)

functional group

amine

SMILES string

CCNC(C)C

InChI

1S/C5H13N/c1-4-6-5(2)3/h5-6H,4H2,1-3H3

InChI key

RIVIDPPYRINTTH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

60.8 °F - closed cup

Flash Point(C)

16 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Giulio Mannocchi et al.
Analytical and bioanalytical chemistry, 412(21), 5125-5145 (2020-02-18)
Interest on keratinized matrix analysis for clinical and forensic purposes has been recently grown due to the wide temporary detection window for psychotropic and toxic substances entrapped after repeated consumption. The aim of this study was the development and full
Chao Zhao et al.
Nature communications, 5, 5079-5079 (2014-10-07)
The remarkable stability of peptide nucleic acids (PNAs) towards enzymatic degradation makes this class of molecules ideal to develop as part of a diagnostic device. Here we report the development of chemically engineered PNAs for the quantitative detection of HIV

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service