Skip to Content
MilliporeSigma
All Photos(2)

Documents

386367

Sigma-Aldrich

Methyl 3,5-dinitrobenzoate

99%

Synonym(s):

3,5-Dinitrobenzoic acid methyl ester, NSC 7317

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(O2N)2C6H3CO2CH3
CAS Number:
Molecular Weight:
226.14
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

mp

107-109 °C (lit.)

SMILES string

COC(=O)c1cc(cc(c1)[N+]([O-])=O)[N+]([O-])=O

InChI

1S/C8H6N2O6/c1-16-8(11)5-2-6(9(12)13)4-7(3-5)10(14)15/h2-4H,1H3

InChI key

POGCCFLNFPIIGW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service