Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

384070

Sigma-Aldrich

2,3,5,6-Tetrafluoro-4-(trifluoromethyl)phenol

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3C6F4OH
CAS Number:
Molecular Weight:
234.07
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

liquid

refractive index

n20/D 1.414 (lit.)

bp

163-164 °C (lit.)

density

1.713 g/mL at 25 °C (lit.)

SMILES string

Oc1c(F)c(F)c(c(F)c1F)C(F)(F)F

InChI

1S/C7HF7O/c8-2-1(7(12,13)14)3(9)5(11)6(15)4(2)10/h15H

InChI key

HZQGKHUTYHEFBT-UHFFFAOYSA-N

General description

2,3,5,6-Tetrafluoro-4-(trifluoromethyl)phenol (Perfluoro-p-cresol), an organofluorine compound, is an aryl fluorinated building block. It is a colorless liquid having hygroscopic properties and a faintly phenolic odor. It has been prepared by using octafluorotoluene as starting reagent. Various physical properties (density, refractive index, freezing point and boiling point) of 2,3,5,6-tetrafluoro-4-(trifluoromethyl)phenol have been reported.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

179.6 °F - closed cup

Flash Point(C)

82 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Knunyants IL and Yakobson GG.
Syntheses of Fluoroorganic Compounds, 171-172 (1985)
Yaws CL.
The Yaws Handbook of Physical Properties for Hydrocarbons and Chemicals, 133-133 (2015)
Kun Qian et al.
Dalton transactions (Cambridge, England : 2003), 47(36), 12759-12768 (2018-08-29)
Dielectric polymer-based nanocomposites have attracted significant attention in recent years for energy storage applications because of their potential high permittivity and breakdown strength. The coupling effect of a nanofiller/matrix interface plays a crucial role in the dielectric and electric properties
[18F]/19F exchange in fluorine containing compounds for potential use in 18F-labelling strategies.
Blom E, et al.
Journal of Labelled Compounds & Radiopharmaceuticals, 52(12), 504-511 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service