Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

380490

Sigma-Aldrich

(2-Iodoethyl)benzene

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2CH2I
CAS Number:
Molecular Weight:
232.06
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.601 (lit.)

bp

122-123 °C/13 mmHg (lit.)

density

1.603 g/mL at 25 °C (lit.)

SMILES string

ICCc1ccccc1

InChI

1S/C8H9I/c9-7-6-8-4-2-1-3-5-8/h1-5H,6-7H2

InChI key

KVTHPKXDLVYNCH-UHFFFAOYSA-N

General description

(2-Iodoethyl)benzene is a halogenated hydrocarbon. It is reported to undergo triethyl borane-mediated intermolecular radical addition with 2H-azirine-3-carboxylate.

Application

(2-Iodoethyl)benzene is suitable for use as starting reagent in the preparation of organic-inorganic hybrid compounds such as, [C6H5CH2NH3]2PbI4, [C6H5CH2CH2SC(NH2 )2]3PbI5 and [C10H7CH2NH3]PbI3. It may be used in the preparation of dioxane-based antiviral agents.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ha Young Kim et al.
Bioorganic & medicinal chemistry letters, 15(13), 3207-3211 (2005-06-02)
Dioxane-based antiviral agents targeted to the hydrophobic binding pocket of Sindbis virus capsid protein were designed by computer graphics molecular modeling and synthesized. Virus production using SIN-IRES-Luc and capsid assembly were monitored to evaluate antiviral activity. A compound with a
Intermolecular alkyl radical addition to methyl 2-(2, 6-dichlorophenyl)-2H-azirine-3-carboxylate.
Lemos A, et al.
Synlett, 10, 1403-1406 (2003)
Preparation and Characterization of [C6H5CH2NH3] 2PbI4,[C6H5CH2CH2SC (NH2) 2] 3PbI5 and [C10H7CH2NH3] PbI3 Organic-Inorganic Hybrid Compounds.
Papavassiliou GC, et al.
Zeitschrift fur Naturforschung, 54b, 1405-1405 (1999)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service