Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

367265

Sigma-Aldrich

Trimethyl(tributylstannyl)silane

97%

Synonym(s):

Tributyl(trimethylsilyl)stannane, (Tributylstannyl)trimethylsilane, (Trimethylsilyl)tributylstannane, (Trimethylsilyl)tributyltin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)3]3SnSi(CH3)3
CAS Number:
Molecular Weight:
363.24
Beilstein:
4127163
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.488 (lit.)

bp

102-103 °C/0.5 mmHg (lit.)

density

1.04 g/mL at 25 °C (lit.)

SMILES string

CCCC[Sn](CCCC)(CCCC)[Si](C)(C)C

InChI

1S/3C4H9.C3H9Si.Sn/c3*1-3-4-2;1-4(2)3;/h3*1,3-4H2,2H3;1-3H3;

InChI key

MZUUBYSIDDKUKE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2 - STOT RE 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service