Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

366587

Sigma-Aldrich

Copper(II) tetrafluoroborate hydrate

Synonym(s):

Copper ditetrafluoroborate hydrate, Cupric tetrafluoroborate hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cu(BF4)2 · xH2O
CAS Number:
Molecular Weight:
237.16 (anhydrous basis)
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

form

solid

composition

Cu(II), 19-22%

concentration

≤25.8% Cu (EDTA titration)

application(s)

battery manufacturing

SMILES string

O.[Cu++].F[B-](F)(F)F.F[B-](F)(F)F

InChI

1S/2BF4.Cu.H2O/c2*2-1(3,4)5;;/h;;;1H2/q2*-1;+2;

InChI key

JYLPBVRGTDBGBM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Copper(II) tetrafluoroborate hydrate may be used as catalyst for chemoselective N-tert-butoxycarbonylation of amines with di-tert-butyl dicarbonate.The product is a starting material for homometallic, trinuclear heteroscorpionate complexes applied in studies of electronic and magnetic properties.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Copper(II) tetrafluoroborate as a novel and highly efficient catalyst for N-tert-butoxycarbonylation of amines under solvent-free conditions at room temperature
Chankeshwara SV, et al.
Tetrahedron Letters, 47(7), 1087-1091 (2006)
Jin Shen et al.
Nature communications, 11(1), 6259-6259 (2020-12-09)
Three-dimensional metal-organic frameworks (MOFs) are cutting-edge materials in the adsorptive removal of trace gases due to the availability of abundant pores with specific chemistry. However, the development of ideal adsorbents combining high adsorption capacity with high selectivity and stability remains
Timothy C. Higgs et al.
Inorganic chemistry, 37(9), 2263-2272 (2001-10-24)
The mixed functionality pyrazole/phenol ligand (2-hydroxyphenyl)bis(pyrazolyl)methane, L1OH, has been used to prepare a series of linear trimetallic systems with the general structural motif [M(3)(L1O)(4)](2+), where M = Mn(2+), Co(2+), Ni(2+), Cu(2+), and Zn(2+). Each of these complexes has been structurally

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service