Skip to Content
MilliporeSigma
All Photos(1)

Documents

358975

Sigma-Aldrich

Ammonium metatungstate hydrate

≥66.5% (W)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(NH4)6H2W12O40 · xH2O
CAS Number:
Molecular Weight:
2956.30 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

form

crystalline

reaction suitability

reagent type: catalyst
core: tungsten

concentration

≥66.5% (W)

mp

100 °C (dec.) (lit.)

SMILES string

N.N.N.N.N.N.O.O[W]1O[W](O)(=O)(O1)O[W]2(=O)(=O)O[W](O)(=O)(O2)O[W]3(=O)(=O)O[W](O)(=O)(O3)O[W]4(=O)(=O)O[W](O)(=O)(O[W]5(=O)(=O)O[W](O)(=O)(O[W]6(=O)(=O)O[W](O)(O)O6)O5)O4

InChI

1S/6H3N.9H2O.32O.12W/h6*1H3;9*1H2;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;/q;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;6*+1;+2/p-8

InChI key

ZBHPUDGBNLMJQH-UHFFFAOYSA-F

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ze-Da Meng et al.
Nanoscale research letters, 6(1), 459-459 (2011-07-22)
WO3-treated fullerene/TiO2 composites (WO3-fullerene/TiO2) were prepared using a sol-gel method. The composite obtained was characterized by BET surface area measurements, X-ray diffraction, scanning electron microscopy, energy dispersive X-ray analysis, transmission electron microscopy, and UV-vis analysis. A methyl orange (MO) solution
Alessandro Chieregato et al.
ChemSusChem, 8(2), 398-406 (2014-12-10)
Glycerol surplus from biodiesel synthesis still represents a major problem in the biofuel production chain. Meanwhile, those in the acrylic acid market are looking for new processes that are able to offer viable alternatives to propylene-based production. Therefore, acrylic acid
Thandavarayan Maiyalagan et al.
Analytical chemistry, 86(15), 7849-7857 (2014-07-16)
Supported tungsten carbide is an efficient and vital nanomaterial for the development of high-performance, sensitive, and selective electrochemical sensors. In this work, tungsten carbide with tube-like nanostructures (WC NTs) supported platinum nanoparticles (PtNPs) are synthesized and explored as an efficient
Luqman Atanda et al.
ChemSusChem, 8(17), 2907-2916 (2015-08-05)
A water-THF biphasic system containing N-methyl-2-pyrrolidone (NMP) was found to enable the efficient synthesis of 5-hydroxymethylfurfural (HMF) from a variety of sugars (simple to complex) using phosphated TiO2 as a catalyst. Fructose and glucose were selectively converted to HMF resulting

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service