Skip to Content
MilliporeSigma
All Photos(3)

Documents

309001

Sigma-Aldrich

1,3,5-Tris(2-hydroxyethyl)isocyanurate

97%

Synonym(s):

1,3,5-Tris(2-hydroxyethyl)-1,3,5-triazine-2,4,6(1H,3H,5H)-trione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H15N3O6
CAS Number:
Molecular Weight:
261.23
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

mp

136-140 °C (lit.)

SMILES string

OCCN1C(=O)N(CCO)C(=O)N(CCO)C1=O

InChI

1S/C9H15N3O6/c13-4-1-10-7(16)11(2-5-14)9(18)12(3-6-15)8(10)17/h13-15H,1-6H2

InChI key

BPXVHIRIPLPOPT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,3,5-Tris(2-hydroxyethyl)isocyanurate (THEIC) is a triazine derivative with three nitrogen atoms in its structure. It can be used as a stabilizer and can be a useful material in applications that require flame-retardancy and water durability.

Application

THEIC can be used to functionalize graphene magnetic nanoparticles to form a catalyst that facilitates the synthesis of benzimidazole compounds. It may also be used in the synthesis of hyperbranched epoxy resin by UV-initiated thiolene click reaction, which finds potential application in the fabrication of insulating materials.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

465.8 °F - open cup

Flash Point(C)

241 °C - open cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Highly efficient preparation of hyperbranched epoxy resins by UV-initiated thiol-ene click reaction
Zhang D, et al.
Progress in Organic Coatings, 101(14), 178-185 (2016)
1, 3, 5-Tris (2-hydroxyethyl) isocyanurate functionalized graphene oxide: a novel and efficient nanocatalyst for the one-pot synthesis of 3, 4-dihydropyrimidin-2 (1 H)-ones
Dekamin MG, et al.
New. J. Chem., 41(14), 6893-6901 (2017)
Rocío Redón et al.
ISRN organic chemistry, 2012, 906839-906839 (2012-01-01)
Amphiphiles with a dendritic structure are attractive materials as they combine the features of dendrimers with the self-assembling properties and interfacial behavior of water-air affinities. We have synthesized three generations of polyester dendrimers and studied their interfacial properties on the
Effects of intumescent flame retardant system consisting of tris (2-hydroxyethyl) isocyanurate and ammonium polyphosphate on the flame retardant properties of high-density polyethylene composites
Khanal S, et al.
Composites Part A: Applied Science and Manufacturing, 112, 444-451 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service