Skip to Content
MilliporeSigma
All Photos(1)

Documents

308137

Sigma-Aldrich

N-Methylpropylamine

96%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2NHCH3
CAS Number:
Molecular Weight:
73.14
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

form

liquid

refractive index

n20/D 1.394 (lit.)

bp

61-63 °C (lit.)

density

0.713 g/mL at 25 °C (lit.)

SMILES string

CCCNC

InChI

1S/C4H11N/c1-3-4-5-2/h5H,3-4H2,1-2H3

InChI key

GVWISOJSERXQBM-UHFFFAOYSA-N

General description

The NMR spectra of N-methylpropylamine coordinated to the paramagnetic 11-tungstocobalto(II)silicate anion (SiW11Co) in dimethylsulfoxide-d6 was studied.

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-25.6 °F - closed cup

Flash Point(C)

-32 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H NMR spectra of some amines coordinated to the paramagnetic polyoxometalate,[SiW 11 Co II O 39] 6
Bull. Korean Chem. Soc., 20, 1145-1148 null
Jyrki Viidanoja
Journal of chromatography. A, 1480, 106-111 (2016-12-21)
A new liquid chromatography-electrospray ionization-tandem Mass Spectrometry (LC-ESI-MS/MS) method was developed for the determination of more than 20 C1-C6 alkyl and alkanolamines in aqueous matrices. The method employs Hydrophilic Interaction Liquid Chromatography Multiple Reaction Monitoring (HILIC-MRM) with a ZIC-pHILIC column

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service