Skip to Content
MilliporeSigma
All Photos(4)

Documents

304468

Sigma-Aldrich

N,N-Dibutylaniline

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5N[(CH2)3CH3]2
CAS Number:
Molecular Weight:
205.34
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Assay

97%

form

liquid

refractive index

n20/D 1.518 (lit.)

bp

269-270 °C (lit.)

density

0.906 g/mL at 25 °C (lit.)

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

CCCCN(CCCC)c1ccccc1

InChI

1S/C14H23N/c1-3-5-12-15(13-6-4-2)14-10-8-7-9-11-14/h7-11H,3-6,12-13H2,1-2H3

InChI key

FZPXKEPZZOEPGX-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yusuke Yamamoto et al.
The Journal of toxicological sciences, 44(9), 585-600 (2019-09-03)
Amino acid derivative reactivity assay (ADRA) has previously been developed as an alternative method to direct peptide reactivity assay (DPRA) to evaluate key event 1 in skin sensitization mechanisms. However, when using alternative methods for skin sensitization, integrated approaches to
Antonella Fontana et al.
Organic & biomolecular chemistry, 13(35), 9214-9222 (2015-08-01)
With the aim to improve the features of surfactant solutions in terms of sustainability and renewability we propose the use of hydrogenated natural and sustainable plant-derived cardanol as an additive to commercial surfactants. In the present study we demonstrated that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service