Skip to Content
MilliporeSigma
All Photos(1)

Documents

295914

Sigma-Aldrich

Methyl difluoroacetate

96%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
F2CHCO2CH3
CAS Number:
Molecular Weight:
110.06
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

refractive index

n20/D 1.334 (lit.)

bp

85-86 °C (lit.)

density

1.26 g/mL at 25 °C (lit.)

SMILES string

COC(=O)C(F)F

InChI

1S/C3H4F2O2/c1-7-3(6)2(4)5/h2H,1H3

InChI key

CSSYKHYGURSRAZ-UHFFFAOYSA-N

General description

The gas-phase reactions of Cl atoms with methyl difluoroacetate have been investigated at 296 ± 2K and atmospheric pressure of synthetic air.

Application

Methyl difluoroacetate has been used:
  • to compose the electrolyte for Lithium ion cells, also showed good inhibition of aluminum cathode current collector corrosion
  • as electrolyte solvent for improvement of thermal stability in lithium-ion batteries

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

65.1 °F - closed cup - (own results)

Flash Point(C)

18.4 °C - closed cup - (own results)

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Methyl difluoroacetate inhibits corrosion of aluminum cathode current collector for lithium ion cells.
Kawamura T, et al.
Electrochemical and Solid-State Letters, 8(9), A459-A463 (2005)
M B Blanco et al.
Environmental science & technology, 44(7), 2354-2359 (2010-03-13)
A 1080 L environmental chamber with in situ FTIR spectroscopy detection was used to study the product distribution and the mechanism of the Cl-initiated photooxidation of a series of fluoroacetates. The gas-phase reactions of Cl atoms with ethyl trifluoroacetate (CF(3)C(O)OCH(2)CH(3))
Properties of carbon anodes and thermal stability in LiPF6/methyl difluoroacetate electrolyte.
Ihara M, et al.
Journal of the Electrochemical Society, 150(11), A1476-A1483 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service