Skip to Content
MilliporeSigma
All Photos(2)

Documents

285072

Sigma-Aldrich

5-Amino-2-naphthalenesulfonic acid

≥95%

Synonym(s):

1-Amino-6-naphthalenesulfonic acid, 1-Amino-6-sulfonaphthalene, 1-Naphthylamine-6-sulfonic acid, 5-Naphthylamine-2-sulfonic acid, 6-Sulfo-1-naphthylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC10H6SO3H
CAS Number:
Molecular Weight:
223.25
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95%

form

solid

solubility

soluble 100 mg/mL, clear, brown (1N NH4OH)

SMILES string

Nc1cccc2cc(ccc12)S(O)(=O)=O

InChI

1S/C10H9NO3S/c11-10-3-1-2-7-6-8(15(12,13)14)4-5-9(7)10/h1-6H,11H2,(H,12,13,14)

InChI key

UWPJYQYRSWYIGZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The crystal structure of 5-amino-2-naphthalenesulfonic acid (1,6-Cleve′s acid) depicted the presence of a sulfonate-aminium group zwitterion.

Application

5-Amino-2-naphthalenesulfonic acid (1,6-Cleve′s acid) was used in the synthesis of nitrate reductase and detection of nitrate reduction by anaerobic bacteria.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P A Wideman et al.
Journal of clinical microbiology, 5(3), 315-319 (1977-03-01)
The laboratory and clinical evaluation of a potassium nitrate-saturated disk for the rapid detection of nitrate reductase production in anaerobes was investigated. The optimal disk concentration and incubation time were determined by utilizing triplicate sets of quadrant plates prepared with
Smith G, et al.
Acta Crystallographica Section E, Structure Reports Online, 60(11), o2014-o2016 (2004)
A S Rodrigues et al.
Chemosphere, 205, 433-442 (2018-05-01)
The electrodegradation of the 4-aminonaphthalene-1-sulfonic acid (4AN1S), 5-aminonaphthalene-2-sulfonic acid (5AN2S) and 8-aminonaphthalene-2-sulfonic acid (8AN2S) was studied, using two electrode materials as anode, BDD and Ti/Pt/PbO2, and two different electrolytes, sodium sulfate and sodium chloride. The highest COD removal rates were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service