Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

264776

Sigma-Aldrich

trans-2-Methyl-2-pentenoic acid

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5CH=C(CH3)CO2H
CAS Number:
Molecular Weight:
114.14
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

>1 (vs air)

Assay

97%

form

liquid

impurities

2% cis-isomer

refractive index

n20/D 1.46 (lit.)

bp

123-125 °C/30 mmHg (lit.)

mp

26-28 °C (lit.)

SMILES string

CC\C=C(/C)C(O)=O

InChI

1S/C6H10O2/c1-3-4-5(2)6(7)8/h4H,3H2,1-2H3,(H,7,8)/b5-4+

InChI key

JJYWRQLLQAKNAD-SNAWJCMRSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Adsorption and desorption of trans-2-methyl-2-pentenoic acid in dichloromethane has been investigated by using in situ attenuated total reflection infrared spectroscopy. Asymmetric hydrogenation of trans-2-methyl-2-pentenoic acid in hexane over two well defined E-40692 and E-5220 Engelhard Pd/Al2O3 catalysts modified with cinchonidine has been studied.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

226.4 °F - closed cup

Flash Point(C)

108 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shuai Tan et al.
Physical chemistry chemical physics : PCCP, 13(43), 19573-19579 (2011-10-06)
Adsorption and desorption of trans-2-methyl-2-pentenoic acid (MPeA) in dichloromethane (CH(2)Cl(2)) were investigated by using in situ attenuated total reflection infrared (ATR-IR) spectroscopy. A liquid flow-through spectroscopic cell allowed for high quality spectra to be obtained from deposited thin films of
Heterogeneous asymmetric hydrogenation and deuteration of 2-methyl-2-pentenoic acid over Pd supported catalysts: proton/deuterium exchange.
Solladie-Cavallo A, et al.
J. Mol. Catal. A: Chem., 195(1), 181-188 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service