Skip to Content
MilliporeSigma
All Photos(1)

Documents

257834

Sigma-Aldrich

Pentacosafluoro-1-iodododecane

97%

Synonym(s):

1-Iodoperfluorododecane, Perfluorododecyl iodide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3(CF2)11I
CAS Number:
Molecular Weight:
745.99
Beilstein:
1718343
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

bp

108-110 °C/18 mmHg (lit.)

mp

95-98 °C (lit.)

SMILES string

FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)I

InChI

1S/C12F25I/c13-1(14,3(17,18)5(21,22)7(25,26)9(29,30)11(33,34)35)2(15,16)4(19,20)6(23,24)8(27,28)10(31,32)12(36,37)38

InChI key

WBYCUETVRCXUPE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Pentacosafluoro-1-iodododecane (1-Iodoperfluorododecane) has been used in E-screen assay to investigate the estrogenic activities of polyfluorinated iodine alkanes in MCF-7 cells.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chang Wang et al.
Environmental health perspectives, 120(1), 119-125 (2011-10-13)
Polyfluorinated iodine alkanes (PFIs) are important intermediates in the synthesis of organic fluoride products. Recently, PFIs have been detected in fluoropolymers as residual raw materials, as well as in the ambient environment. High production volumes and potential environmental releases of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service