Skip to Content
MilliporeSigma
All Photos(2)

Documents

246514

Sigma-Aldrich

Cyclohexyl acetate

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COOC6H11
CAS Number:
Molecular Weight:
142.20
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

liquid

refractive index

n20/D 1.439 (lit.)

bp

172-173 °C (lit.)

density

0.966 g/mL at 25 °C (lit.)

SMILES string

CC(=O)OC1CCCCC1

InChI

1S/C8H14O2/c1-7(9)10-8-5-3-2-4-6-8/h8H,2-6H2,1H3

InChI key

YYLLIJHXUHJATK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The dermatologic and toxicologic review of cyclohexyl acetate, used as a fragrance material, was presented.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

136.4 °F - DIN 51758

Flash Point(C)

58 °C - DIN 51758

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S P Bhatia et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 46 Suppl 12, S52-S55 (2008-10-11)
A toxicologic and dermatologic review of cyclohexyl acetate when used as a fragrance ingredient is presented.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service