Skip to Content
MilliporeSigma
All Photos(1)

Documents

245259

Sigma-Aldrich

Dicarbonylcyclopentadienyl cobalt(I)

technical grade

Synonym(s):

CpCo(CO)2, Cobalt dicarbonyl cyclopentadiene, Cyclopentadienyl cobalt(I) dicarbonyl, Cyclopentadienylcobalt dicarbonyl, Cyclopentadienyldicarbonylcobalt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C5H5Co(CO)2
CAS Number:
Molecular Weight:
180.05
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:

grade

technical grade

form

liquid

reaction suitability

core: cobalt
reagent type: catalyst

bp

139-140 °C/710 mmHg (lit.)

storage temp.

2-8°C

SMILES string

[Co].[C-]#[O+].[C-]#[O+].[CH]1[CH][CH][CH][CH]1

InChI

1S/C5H5.2CO.Co/c1-2-4-5-3-1;2*1-2;/h1-5H;;;

InChI key

HUSLGLMTBHLWKG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Catalyst for the cyclotrimerization of alkynes.
Mediates the [2+2+2]-cyclization of allenediynes to tricyclic systems.

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Carc. 2 - Flam. Liq. 3 - Muta. 2 - Repr. 2 - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

80.6 °F - closed cup

Flash Point(C)

27 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetrahedron, 62, 10582-10582 (2006)
Room Temperature CpCo-Mediated Cyclization of alpha,delta,omega-Enediynes to Rearranging Strained Tricylic Dienes. Some Observations of Kinetic versus Thermodynamic Control.
J. Kevin Cammack et al.
The Journal of organic chemistry, 61(14), 4798-4800 (1996-07-12)
Dmitri V Konarev et al.
Dalton transactions (Cambridge, England : 2003), 49(46), 16821-16829 (2020-11-13)
The reduction of copper(ii) octafluoro- {CuII(F8Pc)} and hexadecafluorophthalocyanines {CuII(F16Pc)} by NaCpCo(CO)2 in the presence of cryptand[2.2.2] yields new crystalline {cryptand(Na+)}[CuII(F8Pc)˙3-]-·2C6H4Cl2 (1) and {cryptand(Na+)}2[CuII(F16Pc)4-]2-·C6H14 (2) salts. Together with two previously characterized salts of CuII(FxPc) (x = 8 and 16), this allows

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service