Skip to Content
MilliporeSigma
All Photos(1)

Documents

227447

Sigma-Aldrich

4,9-Dioxa-1,12-dodecanediamine

99%

Synonym(s):

1,4-Bis(3-aminopropoxy)butane, 4,9-Dioxadodecanediamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2N(CH2)3O(CH2)4O(CH2)3NH2
CAS Number:
Molecular Weight:
204.31
Beilstein:
1750788
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

99%

form

liquid

refractive index

n20/D 1.4609 (lit.)

bp

134-136 °C/4 mmHg (lit.)

density

0.962 g/mL at 25 °C (lit.)

SMILES string

NCCCOCCCCOCCCN

InChI

1S/C10H24N2O2/c11-5-3-9-13-7-1-2-8-14-10-4-6-12/h1-12H2

InChI key

YOOSAIJKYCBPFW-UHFFFAOYSA-N

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1A

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nikolay Tamkovich et al.
Bioorganic & medicinal chemistry, 24(6), 1346-1355 (2016-02-24)
A novel series of metal-free artificial ribonucleases (aRNases) was designed, synthesized and assessed in terms of ribonuclease activity and ability to inactivate influenza virus WSN/A33/H1N1 in vitro. The compounds were built of two short peptide fragments, which include Lys, Ser

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service